Anti-SP Rabbit Monoclonal Antibody [clone: EPR3959]

Supplier: GeneTex
GTX62656
89364-552EA 694.85 CAD
89364-552
Anti-SP Rabbit Monoclonal Antibody [clone: EPR3959]
Antibodies
Rabbit monoclonal antibody [EPR3959] to Substance P

IHC-P: 1/250 - 1/500. Perform heat mediated antigen retrieval with citrate buffer pH 6 before commencing with IHC staining protocol. Is unsuitable for ICC/IF.

Type: Primary
Antigen: SP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR