Anti-SUV39H2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX25264
89361-606EA 882 CAD
89361-606
Anti-SUV39H2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SUV39H2

No signal obtained yet but low background observed in preliminary tests on Human Testis extracts at up to 1µg/ml. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?

Type: Primary
Antigen: SUV39H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR