Anti-SWC9 Mouse Monoclonal Antibody

GTX75669
89367-910EA 647.22 CAD
89367-910
Anti-SWC9 Mouse Monoclonal Antibody
SWC9 is a cell surface antigen expressed on mature pig macrophages. Freshly isolated blood monocytes are SWC9 negative but do express SWC9 after culturing. SWC9 is also weakly expressed on porcine thymocytes.

Type: Primary
Antigen: SWC9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR