Anti-TAG72 Mouse Monoclonal Antibody [clone: B72.3]

Supplier: GeneTex
GTX21320
89351-632EA 915.67 CAD
89351-632
Anti-TAG72 Mouse Monoclonal Antibody [clone: B72.3]
Antibodies
The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for TAG 72. About 60% of carcinoma patients express TAG 72 in their sera.

Type: Primary
Antigen: TAG72
Clonality: Monoclonal
Clone: B72.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR