Anti-TAG72 Mouse Monoclonal Antibody

GTX73796
89353-278EA 772.59 CAD
89353-278
Anti-TAG72 Mouse Monoclonal Antibody
This antibody is specific to an oncofetal antigen of >1,000 kD known as tumor associated glycoprotein (TAG-72) or CA 72-4. This antibody has been shown to be distinct from B72.3 by reciprocal competition radio-immunoassays and it has a Ka higher than that of B72.3. TAG-72 is expressed in colon, breast, pancreatic, ovarian, lung and gastric cancer.

Type: Primary
Antigen: TAG72
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR