Anti-Topoisomerase Ii Beta Rabbit Polyclonal Antibody

GTX15565
89351-192EA 460.62 CAD
89351-192
Anti-Topoisomerase Ii Beta Rabbit Polyclonal Antibody
Topoisomerase II beta plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is present ubiquitously in normal cells and is upregulated in tumors and proliferating cells. Topoisomerase II beta is also implicated in drug resistance of tumour cells.

Type: Primary
Antigen: Topoisomerase Ii Beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR