Anti-Trophoblast Mouse Monoclonal Antibody [clone: NDOG1]

GTX11460
89358-088EA 456.78 CAD
89358-088
Anti-Trophoblast Mouse Monoclonal Antibody [clone: NDOG1]
This protein, can be isolated from bovine and ovine sources. TP-1 is a member of the type-1 interferon class, is 172 amino acids in length, and is related to human IFN-omega. The sequences of the bovine (22 and 24 kDa) and the ovine protein (17 kDa) are 85 percent identical and about 79 percent identical with that for bovine IFN-alpha-2.

Type: Primary
Antigen: Trophoblast
Clonality: Monoclonal
Clone: [NDOG1]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR