Anti-TDT Mouse Monoclonal Antibody [clone: SPM149]

GTX15242
89351-030EA 621.72 CAD
89351-030
Anti-TDT Mouse Monoclonal Antibody [clone: SPM149]
TdT is a nuclear enzyme involved in the recombination of Immunoglobulin and T cell receptor genes. TdT is expressed in early lymphoid cells and by tumour cells in certain classes of acute lymphoid leukaemia.

Type: Primary
Antigen: TDT
Clonality: Monoclonal
Clone: SPM149
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR