Anti-TENASCIN-C Rabbit Monoclonal Antibody

GTX62552
89364-502EA 556.26 CAD
89364-502
Anti-TENASCIN-C Rabbit Monoclonal Antibody
Tenascin-C is the founding member of a family of extracellular matrix glycoproteins that includes tenascin-X, -R and -W. Tenascin-C interacts with several other extracellular matrix molecules and cell-surface receptors, thus affecting tissue architecture, tissue resilience and cell responses . It also modulates cell migration; proliferation; and cellular signaling through induction of pro-inflammatory cytokines and oncogenic signaling molecules, amongst other mechanisms . Tenascin-C is highly expressed during embryonic development, tissue repair and in pathological situations such as chronic inflammation and cancer . In the adult, Tenascin-C remains present in tendons and myotendinous junctions in the perichondrium and periosteum, as well as in smooth muscle .

Type: Primary
Antigen: TENASCIN-C
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR