Anti-Tetraspanin-8 Chicken Polyclonal Antibody

GTX82454
89368-086EA 666.36 CAD
89368-086
Anti-Tetraspanin-8 Chicken Polyclonal Antibody
The protein encoded by this gene is a member of the transmembrane 4 superfamily. also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development. activation. growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins. This protein is expressed in different carcinomas. The use of alternate polyadenylation sites has been found for this gene.

Type: Primary
Antigen: Tetraspanin-8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR