Anti-TFF1 Mouse Monoclonal Antibody

GTX17829
89351-398EA 494.72 CAD
89351-398
Anti-TFF1 Mouse Monoclonal Antibody
This protein was identified by differential screening of cDNA libraries from estrogen responsive breast cancer cell lines. It is expressed in normal gastric and small intestinal mucosa and breast epithelium. In cancer, this protein is expressed in gastric carcinoma and gynecological cancers. Presence of this protein in breast carcinomas provides research investigators supplimentary information for identifying patients with likely positive response to hormone therapy and a favorable prognosis.

Type: Primary
Antigen: TFF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR