Anti-TG Mouse Monoclonal Antibody [clone: 5E6]

Supplier: GeneTex
GTX21982
89351-722EA 933.32 CAD
89351-722
Anti-TG Mouse Monoclonal Antibody [clone: 5E6]
Antibodies
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3.Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimotois thyroiditis, Gravesi diseases), with the highest level observed in Hashimotois thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.

ELISA: Use at an assay dependent dilution. Can be used in an ELISA pair with GTX21984 as coating. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: 5E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR