Anti-TLR1 Mouse Monoclonal Antibody [clone: GD2.F4]

Supplier: GeneTex
GTX11209
89356-976EA 833.73 CAD
89356-976
Anti-TLR1 Mouse Monoclonal Antibody [clone: GD2.F4]
Antibodies
Mouse monoclonal antibody [GD2.F4] to TLR1

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/100. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TLR1
Clonality: Monoclonal
Clone: GD2.F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR