Anti-TLR9 Mouse Monoclonal Antibody [clone: 5G5]

Supplier: GeneTex

GTX76032
89367-944EA 622.13 CAD
89367-944
Anti-TLR9 Mouse Monoclonal Antibody [clone: 5G5]
Antibodies
Mouse monoclonal antibody [5G5] to TLR9

Recommended Starting Dilutions: Western Blotting: 1/50 - 1/200 Flow Cytometry: 1/10 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR