Anti-TNFA Mouse Monoclonal Antibody

GTX74959
89365-646EA 584.7 CAD
89365-646
Anti-TNFA Mouse Monoclonal Antibody
Recognises human TNF alpha. Clone 4C6-H6 has been reported to work in Western Blotting.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR