Anti-TNFR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15564
89351-190EA 587.54 CAD
89351-190
Anti-TNFR2 Rabbit Polyclonal Antibody
Tumor Necrosis Factor (TNF) is a cytokine whose function is mediated through two distinct cell surface receptors (TNFR 1 and TNFR 2) that are included in the TNF receptor superfamily along with FAS antigen and CD40. Tumor necrosis factor receptor 1 and 2 (TNFR 1 and TNFR 2) are 55 and 75 kDa members of a family cell surface molecules including nerve growth factor receptor, Fas/Apo1, CD30, Ox40, and 4-1BB, which are characterized by cysteine rich motifs in the extracellular domain.

IHC-P: Working dilution: Ready-to-use for 30 min at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR