Anti-TNFAIP1 Chicken Polyclonal Antibody

GTX82245
89368-060EA 642.96 CAD
89368-060
Anti-TNFAIP1 Chicken Polyclonal Antibody
Chicken polyclonal to Tumor necrosis factor. alpha-induced protein 1. endothelial

ELISA (Direct-only tested against protein): Affi-pure IgY as primary antibody (1 ug/ml) and Goat anti-IgY HRP as 2nd antibody. WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: TNFAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR