Anti-TNFRSF14 Rabbit Polyclonal Antibody

GTX113588
89357-630EA 258.54 CAD
89357-630
Anti-TNFRSF14 Rabbit Polyclonal Antibody
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to this receptor protein has been shown to be part of the viral entry mechanism. The cytoplasmic region of this receptor was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. [provided by RefSeq]

Type: Primary
Antigen: TNFRSF14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR