Anti-TPPP Rabbit Monoclonal Antibody

GTX62320
89364-396EA 556.26 CAD
89364-396
Anti-TPPP Rabbit Monoclonal Antibody
Tubulin polymerization-promoting protein (TPPP) is an unfolded brain-specific protein that interacts with the tubulin/microtubule system, and is enriched in human pathological brain inclusions . TPPP can induce aggregation of alpha-synuclein, and can accumulate in oligodendroglial cell bodies containing fibrillized alpha-synuclein multiple system atrophy (MSA), a neurodegenerative disease. TPPP is a constituent of myelin and a high-affinity ligand for myelin basic protein (MBP), and has been shown to colocalize with MBP in myelin in normal human brains .

Type: Primary
Antigen: TPPP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR