Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R]

Supplier: GeneTex
GTX48033
89363-642EA 601.95 CAD
89363-642
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R]
Mouse Monoclonal antibody to TRA-1-60

For flow cytometry, we recommend using 0.5-1 ug per million cells. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRA-1-60
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR