Anti-TRAF4 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77728
89366-266EA 646.08 CAD
89366-266
Anti-TRAF4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to TRAF-4

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: TRAF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR