Anti-TRAPPC9 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX48542
89363-662EA 963.48 CAD
89363-662
Anti-TRAPPC9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TRAPPC9

WB: 1:100-1:2000

ICC/IF: 1:10-1:2000

IP: 1:10-1:500

Type: Primary
Antigen: TRAPPC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR