Anti-TRK B Rabbit Monoclonal Antibody

GTX61736
89364-152EA 556.26 CAD
89364-152
Anti-TRK B Rabbit Monoclonal Antibody
There is increasing evidence that neurotrophins and their receptors play an important role in regulating development of both the central and the peripheral nervous systems . The binding of neurotrophin ligands to their respective Trk cellular receptors initiates intracellular signals essential for the growth and survival of neurons. The site of neurotrophin binding has been located to the fifth extracellular domain of the Trk receptor, with this region regulating both the affinity and specificity of Trk receptor:neurotrophin interaction . The TrkB receptor tyrosine kinase and its ligand, BDNF, have an essential role in certain forms of synaptic plasticity. Results provide evidence that TrkB mediates hippocampal plasticity via recruitment of PLCgamma, and by subsequent phosphorylation of CaMKIV . Also, TrkB has been found to be essential to the development of GABAergic neurons and regulates synapse formation in addition to its role in the development of axon terminals .

Type: Primary
Antigen: TRK B (Phospho TYR817)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR