Anti-TRPE Mouse Monoclonal Antibody

Supplier: GeneTex

GTX16153
89367-460EA 795.87 CAD
89367-460
Anti-TRPE Mouse Monoclonal Antibody
Antibodies
This antibody can be used for analysis of expression of proteins alone or in conjunction with protein specific antibodies. Can also be used as a negative control antibody.

Type: Primary
Antigen: TRPE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR