Anti-TRP-P8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111818
89356-862EA 881.1 CAD
89356-862
Anti-TRP-P8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TRPM8 (transient receptor potential cation channel, subfamily M, member 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRP-P8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR