Anti-TSC2 Rabbit Monoclonal Antibody

GTX61245
89363-896EA 556.26 CAD
89363-896
Anti-TSC2 Rabbit Monoclonal Antibody
Tuberous sclerosis complex-2 (TSC2 also known as Tuberin) is a tumor suppressor that forms a complex with TSC1 (Hamartin ) and this complex is known to control various cellular functions including cell cycle, endocytosis, adhesion, and transcription . The C-terminal region of TSC2 contains a GTPase-activating protein (GAP) domain which interacts with Rap1, Rab5 and Rheb . The TSC1/TSC2 complex inhibits phosphorylation of S6kinase and 4E-BP1 through inactivation of mTOR . Additionally, binding of 14-3-3 beta to TSC2 at Ser1210 reduce the ability of the complex to inhibit phosphorylation of S6 kinase . Phosphorylation by ATK at Ser924 and Thr1518 inactivates TSC2 and disrupts its interaction with TSC1 . Tuberous sclerosis (TSC), an autosomal dominant disorder that affects 1 in 6000 individuals, is caused by a mutation in either the TSC1 or TSC2 tumor suppressor gene.

Type: Primary
Antigen: TSC2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR