Anti-TSLP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX85059
89353-914EA 694.17 CAD
89353-914
Anti-TSLP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TSLP

Type: Primary
Antigen: TSLP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR