Anti-Tubulin beta III Mouse Monoclonal Antibody

Supplier: GeneTex
GTX78442
89353-846EA 933.78 CAD
89353-846
Anti-Tubulin beta III Mouse Monoclonal Antibody
Antibodies
The betaIII-tubulin isotype is present dominantly in cells of neuronal origin and it is one of the earliest marker of neuronal differentiation. Class III beta-tubulin, is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The neuron-associated class III beta-tubulin isotype is most abundant in cells of neuronal origin but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.

Type: Primary
Antigen: Tubulin beta III
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR