Anti-U1 SNRNP70 Rabbit Polyclonal Antibody

GTX101864
89349-314EA 276.98 CAD
89349-314
Anti-U1 SNRNP70 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to U1 snRNP70 (small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: U1 SNRNP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR