Anti-UBC9 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX101514
89349-090EA 248.06 CAD
89349-090
Anti-UBC9 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to UBE2I (ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: UBC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR