Anti-UBTF Rabbit Monoclonal Antibody

GTX61982
89364-252EA 556.26 CAD
89364-252
Anti-UBTF Rabbit Monoclonal Antibody
The eukaryotic upstream binding factor (UBF), recognizes the ribosomal RNA gene promoter and activates transcription mediated by RNA polymerase I through cooperative interactions with the species-specific factor, SL1. UBF binds specifically to the upstream control element and core of the rRNA gene promoter to activate transcription in a binding site-dependent manner . Experiments reveal that a cooperative interaction between UBF and SL1 leads to the formation of a new protein-DNA complex at the UCE and core elements which indicate that formation of the UBF-SL1 complex is vital for transcriptional activation by UBF . UBF contains six HMG box domains. The contribution of each HMG box motif to its function is different . Its believed that HMG motifs are the DNA-binding domains of UBF. Altering one of these boxes, as in the second form of UBF, may alter the functional characteristics of the transcription factor. Thus, the existence of different forms of UBF may have important ramifications for transcription by RNA polymerase I

Type: Primary
Antigen: UBTF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR