Anti-UCP2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89094
89366-976EA 919.07 CAD
89366-976
Anti-UCP2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Uncoupling protein 2 / UCP2

Approx 35kDa band observed in Human Liver lysates (calculated MW of 33.2kDa according to NP_003346.2). Recommended concentration: 0.2-1µg/ml..

Type: Primary
Antigen: UCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR