Anti-Avpr1b Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX78015
89366-480EA 742.55 CAD
89366-480
Anti-Avpr1b Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Vasopressin V1B Receptor

For IHC-P: Use at a concentration of 4 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: Avpr1b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR