Anti-VCAM1 Rat Monoclonal Antibody [clone: M/K-2]

Supplier: GeneTex

GTX19569
89360-644EA 609.23 CAD
89360-644
Anti-VCAM1 Rat Monoclonal Antibody [clone: M/K-2]
Antibodies
VCAM1 is important in cell-cell recognition. Appears to function in leukocyte-endothelial cell adhesion. Interacts with the beta 1 integrin VLA4 on leukocytes, and mediates both adhesion and signal transduction. The VCAM1/VLA4 interaction may play a pathophysiologic role both in immune responses and in leukocyte emigration to sites of inflammation.

IF: Use at an assay dependent dilution. IHC-Fr: (Not suitable for use with parafin sections) Use at an assay dependent dilution. FuncS: Use at an assay dependent dilution. Studies of cell-cell interaction. Cell adhesion assays. Inhibition of lymphopoiesis in Whitlock-Witte long term marrow cultures. Immunoprecipitation of the VCAM1 integrin complex. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR