Anti-KDR Rabbit Polyclonal Antibody

Supplier: Genetex
GTX25473
89361-648EA 925.15 CAD
89361-648
Anti-KDR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to VEGFR-2 ( phospho Y1054 + Y1059 )

WB: Use at a concentration of 0.1 - 1 ?g/ml. Detects a band of approximately 200 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR