Anti-FLT1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX30653
89367-096EA 925.6 CAD
89367-096
Anti-FLT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to VEGFR-1 + VEGFR-2

This antibody is useful for Western blot analysis, where a band is seen at ~150 kDa (the theoretical molecular weight of mouse VEGFR1 is ~150 kDa and human VEGFR1 is ~151 kDa). An unknown band is also seen at ~50 kDa.Suggested Starting Dilution:*Western Blot: 1:250-1:1,000 (ECL)*Optimal working dilutions should be determined by the investigator.

Type: Primary
Antigen: Flt1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR