Anti-VIL1 Rabbit Monoclonal Antibody

GTX63140
89364-634EA 585.93 CAD
89364-634
Anti-VIL1 Rabbit Monoclonal Antibody
Villin-1 is a member of a family of calcium-regulated actin-binding proteins. This protein represents a dominant part of the brush border cytoskeleton which functions in the capping, severing, and bundling of actin filaments. Villin-1 is a very early marker of cells involved in absorption and its expression is highly increased during intestinal cell differentiation.

Type: Primary
Antigen: VIL1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR