Anti-VDR Goat Polyclonal Antibody

Supplier: GeneTex
GTX89509
89366-990EA 919.07 CAD
89366-990
Anti-VDR Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to VDR

Approx 40-45kDa band observed in human brain lysates (calculated MW of 48.3kDa according to NP_000367.1and NP_001017535.1. Recommended concentration: 0.3-1.0µg/ml. Both transcript variants (NP_000367; NP_001017535) encode the same protein.

Type: Primary
Antigen: VDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR