Anti-VTN Rabbit Monoclonal Antibody

GTX61399
89363-966EA 556.26 CAD
89363-966
Anti-VTN Rabbit Monoclonal Antibody
Vitronectin, a 75 kDa polypeptide along with a 10 kDa fragment, is a glycoprotein present on most cell surfaces, in extracellular fluids and plasma . Vitronectin promotes cellular adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway and binds to several serpin serine protease inhibitors . As a member of the integrin family, Vitronectin contains the cell binding sequence arg gly asp (RGD), first found in fibronectin, and serves as a cell-to-substrate adhesion molecule . Vitronectin acts as an inhibitor of the complement cascade by binding to the C5b9 complex and it has also been shown to have has a novel and direct role in self-senescent-self intercellular recognition leading to macrophage phagocytosis of cells undergoing apoptosis .

Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR