Anti-VP16 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30776
89362-996EA 900.02 CAD
89362-996
Anti-VP16 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR