Anti-WAPL Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX115664
89358-380EA 881.1 CAD
89358-380
Anti-WAPL Rabbit Polyclonal Antibody
Antibodies
May play a role in cell growth.

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: WAPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR