Anti-XPF Mouse Monoclonal Antibody [clone: 51]

GTX23299
89351-970EA 565.28 CAD
89351-970
Anti-XPF Mouse Monoclonal Antibody [clone: 51]
Mouse monoclonal [51] to XPF

Western blotting: use at a concentration of 1 - 2?g / ml for 2hrs at RT. This antibody is not suitable for Immunohistochemistry. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of ~ 110 kDa, which corresponds to the predicted molecular weight of XPF. This antibody is not suitable for immunohistochemistry.

Type: Primary
Antigen: XPF
Clonality: Monoclonal
Clone: 51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR