Anti-XRCC4 Rabbit Polyclonal Antibody

GTX70263
89364-832EA 400.16 CAD
89364-832
Anti-XRCC4 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to XRCC4

  • Recommended Starting Dilutions: Western Blotting: Max Dilution 1/2000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: XRCC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR