Anti-ZO1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108592
89355-604EA 710.73 CAD
89355-604
Anti-ZO1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZO-1 (tight junction protein 1 (zona occludens 1))

Suggested starting dilutions are as follows: WB: 1:500-1:3000[The use of GTX108592 for mouse is based on IHC-P testing. For WB using mouse samples, we recommend GTX108613], IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ZO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR