ZetexPlus Heat Resistant Gloves, Newtex Industries
2100014 2100011 2100012
32885-383PR 270.09 CAD
32885-383 32885-348 89135-682
ZetexPlus Heat Resistant Gloves, Newtex Industries
Protection from temperatures up to 1093°C (2000°F)
Common Applications: Foundries and Casting Operations, Welding and Hot Works Operations, Glass and Ceramic Plants Production, Power Generation, Petrochemical and Refining, Laboratory and Testing Facilities.
Woven from 100% premium ZetexPlus texturized fiberglass and treated with a proprietary vermiculite coating. Double Stitched and reinforced. Premium wool liner. Available in 28, 35.6 or 58.4cm (11, 14, or 23") lengths. 35.6cm (14") cuff protects wrists. 58.4cm (23") glove features a gauntlet style cuff. One size fits all. Made in the USA.
Certifications: EN 388, EN 407