NXP 150 Z-Flex Aluminized Heat Resistant Apron, Newtex Industries
2100061 2100060 2100062
30617-972EA 146.93 CAD
30617-972 30617-974 30617-970
NXP 150 Z-Flex Aluminized Heat Resistant Apron, Newtex Industries
Protection from short duration ambient heat up to 93°C (200°F) and radiant heat up to 1093°C (2000°F)
Common Applications: Foundries and Casting Operations, Welding and Hot Works Operations, Glass and Ceramic Plants Production, Power Generation, Petrochemical and Refining, Laboratory and Testing Facilities.
Made from Z-Flex Multilayer Aluminized technology, which reflects 95% of radiant heat. Available in 3 sizes: Small (24” wide x 30” long), Medium (24” wide x 36” long), and Large (29” wide x 42” long). Made in the USA.
Certifications: Z-Flex materials have been tested in accordance with ASTM F955, ASTM F1939, EN 532, EN 367, EN 366, EN 407, and MIL-C-24929.