NXP 150 Z-Flex Aluminized Heat Resistant Coat, Newtex Industries
Supplier: Newtex
2100078
30617-962EA 825.17 CAD
30617-962
NXP 150 Z-Flex Aluminized Heat Resistant Coat, Newtex Industries
Protection from short duration ambient heat up to 93°C (200°F) and radiant heat up to 1093°C (2000°F)
Common Applications: Foundries and Casting Operations, Welding and Hot Works Operations, Glass and Ceramic Plants Production, Power Generation, Petrochemical and Refining, Laboratory and Testing Facilities.
Made from Z-Flex Multilayer Aluminized technology, which reflects 95% of radiant heat. Fire resistant cotton inner lining. Fabricated without bulk insulation layers to allow for flexibility and freedom of movement. Measurements: Chest fits 101–119cm (41–47"), waist fits 94–122cm (37–48"), inseam is 74cm (29"). Made in the USA.
Certifications: Z-Flex materials have been tested in accordance with ASTM F955, ASTM F1939, EN 532, EN 367, EN 366, EN 407, and MIL-C-24929.