N-Acetonylpyridinium chloride ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Acetonylpyridiniumchloride

A0828-5G
TCA0828-005GEA 142.2 CAD
TCA0828-005G
N-Acetonylpyridinium chloride ≥99.0% (by HPLC, titration analysis)
N-Acetonylpyridinium chloride

CAS Number: 42508-60-1
MDL Number: MFCD00031949
Molecular Formula: C8H10ClNO
Molecular Weight: 171.62
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207

https://ca.vwr.com/stibo/web/std.lang.all/85/70/28928570.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR