TCIAA1146

A1146-10G A1146-25G
TCA1146-25GEA 271.4 CAD
TCA1146-010G TCA1146-25G
TCIAA1146
stabilized with MEHQ
CAS Number: 2160-89-6
Molecular Formula: C6H4F6O2
Molecular Weight: 222.09
Form: Liquid
Color: Colorless

Product Categories: Materials Chemistry, Polymer Research Reagents, Reagent for High-Performance Polymer Research
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR