2-Amino-4-methylbenzothiazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1344-25G
TCA1344-25GEA
145.36
CAD
TCA1344-25G
2-Amino-4-methylbenzothiazole ≥98.0% (by titrimetric analysis)
2-Amino-4-methylbenzothiazole
CAS Number: 1477-42-5
MDL Number: MFCD00005793
Molecular Formula: C8H8N2S
Molecular Weight: 164.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 137
Formula:
C₈H₈N₂S Melting Pt: 137 °C |
MDL Number:
MFCD00005793 CAS Number: 1477-42-5 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...